Product Details

SNP ID
rs148193888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48750241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGCATTAATGCCCACCCACTC[A/G]GGCAGGAAGGCCGCCTCCGGCTTAA
Phenotype
MIM: 609436 MIM: 211100 MIM: 609278 MIM: 609623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF21 PubMed Links

Gene Details

Gene
FGF21
Gene Name
fibroblast growth factor 21
There are no transcripts associated with this gene.

Gene
FUT1
Gene Name
fucosyltransferase 1 (H blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000148.3 2016 Silent Mutation CCC,CCT P347P NP_000139.1
XM_006723127.2 2016 Silent Mutation CCC,CCT P470P XP_006723190.1
XM_017026552.1 2016 Silent Mutation CCC,CCT P470P XP_016882041.1
XM_017026553.1 2016 Silent Mutation CCC,CCT P347P XP_016882042.1
Gene
IZUMO1
Gene Name
izumo sperm-egg fusion 1
There are no transcripts associated with this gene.

Gene
RASIP1
Gene Name
Ras interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details