Product Details

SNP ID
rs148194580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43612510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACACCTGGCAGAAGGGGAAGCC[A/G]CAGCTCCAAGAGGTCACCCAGCAGG
Phenotype
MIM: 609744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
SRRM5
Gene Name
serine/arginine repetitive matrix 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145641.1 410 Missense Mutation CAC,CGC H130R NP_001139113.1
Gene
ZNF428
Gene Name
zinc finger protein 428
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182498.3 410 Intron NP_872304.2

View Full Product Details