Product Details

SNP ID
rs148246051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50476229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCCCCGTCCGACGGGAGCGGAC[C/G]GAGGAGCAGAGGGAGAAAGCGAGGG
Phenotype
MIM: 614545 MIM: 160793
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175063.5 175 Intron NP_778233.4
NM_206538.3 175 Intron NP_996261.1
XM_006723163.3 175 Intron XP_006723226.1
XM_011526818.2 175 Intron XP_011525120.1
Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 175 Missense Mutation CGT,GGT R54G NP_001295358.1
NM_138411.2 175 Missense Mutation CGT,GGT R54G NP_612420.1
XM_011526405.2 175 Missense Mutation CGT,GGT R54G XP_011524707.2
XM_011526406.2 175 Missense Mutation CGT,GGT R54G XP_011524708.2
Gene
MYBPC2
Gene Name
myosin binding protein C, fast type
There are no transcripts associated with this gene.

View Full Product Details