Product Details

SNP ID
rs148350115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:362411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCGGTCAGGAACGCACTTGTCC[A/G]ACTGAGCTTTGGGCCCTAGTGGGGG
Phenotype
MIM: 609503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THEG PubMed Links

Gene Details

Gene
THEG
Gene Name
theg spermatid protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016585.4 1674 Missense Mutation NP_057669.1
NM_199202.2 1674 Missense Mutation NP_954672.1
XM_011528049.2 1674 Missense Mutation XP_011526351.1
XM_011528050.2 1674 Missense Mutation XP_011526352.1
XM_011528051.2 1674 Missense Mutation XP_011526353.1
XM_011528052.2 1674 Intron XP_011526354.1

View Full Product Details