Product Details

SNP ID
rs148520728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48461988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCGTGGACGTGCGCTGGCGCTG[C/G]ATGTGCCTGCTCTTCTCCTGCTCCT
Phenotype
MIM: 602488 MIM: 610597 MIM: 603953
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYTH2 PubMed Links

Gene Details

Gene
CYTH2
Gene Name
cytohesin 2
There are no transcripts associated with this gene.

Gene
GRWD1
Gene Name
glutamate rich WD repeat containing 1
There are no transcripts associated with this gene.

Gene
KCNJ14
Gene Name
potassium voltage-gated channel subfamily J member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013348.3 471 Missense Mutation TGC,TGG C88W NP_037480.1
Gene
LOC105372430
Gene Name
uncharacterized LOC105372430
There are no transcripts associated with this gene.

View Full Product Details