Product Details

SNP ID
rs148567057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42006737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCGGCCGACTCCACAGGCACCT[C/T]CATGCGCTGCACGGTGAGGAAGGCG
Phenotype
MIM: 600283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIK5 PubMed Links

Gene Details

Gene
GRIK5
Gene Name
glutamate ionotropic receptor kainate type subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301030.1 3397 Missense Mutation AAG,GAG K649E NP_001287959.1
NM_002088.4 3397 Missense Mutation AAG,GAG K649E NP_002079.3
XM_005258821.3 3397 Missense Mutation AAG,GAG K649E XP_005258878.1
XM_011526862.2 3397 Missense Mutation AAG,GAG K650E XP_011525164.1
XM_011526863.2 3397 Missense Mutation AAG,GAG K649E XP_011525165.1
XM_011526865.1 3397 Missense Mutation AAG,GAG K650E XP_011525167.1
XM_011526867.1 3397 Missense Mutation AAG,GAG K581E XP_011525169.1
XM_011526868.1 3397 Missense Mutation AAG,GAG K569E XP_011525170.1
XM_011526869.2 3397 Intron XP_011525171.1
XM_011526870.2 3397 Intron XP_011525172.1
XM_011526871.2 3397 Intron XP_011525173.1
XM_017026713.1 3397 Missense Mutation AAG,GAG K582E XP_016882202.1

View Full Product Details