Product Details

SNP ID
rs148630931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35643623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGACTGTACCACCTCCTGGAA[C/T]CCGGGGCTGCATGCGGGTGGCACCA
Phenotype
MIM: 124089 MIM: 609358 MIM: 613232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX6B1 PubMed Links

Gene Details

Gene
COX6B1
Gene Name
cytochrome c oxidase subunit 6B1
There are no transcripts associated with this gene.

Gene
ETV2
Gene Name
ETS variant 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300974.1 1513 Silent Mutation AAC,AAT N102N NP_001287903.1
NM_001304549.1 1513 Intron NP_001291478.1
NM_014209.3 1513 Silent Mutation AAC,AAT N195N NP_055024.2
XM_005258652.2 1513 Silent Mutation AAC,AAT N223N XP_005258709.1
XM_011526624.2 1513 Silent Mutation AAC,AAT N102N XP_011524926.1
XM_017026472.1 1513 Silent Mutation AAC,AAT N196N XP_016881961.1
Gene
RBM42
Gene Name
RNA binding motif protein 42
There are no transcripts associated with this gene.

View Full Product Details