Product Details
- SNP ID
-
rs148666375
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37235564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGTTTCAGGGATCAGTGATGTTC[A/G]GTGATGTGTCCATAGACTTCTCTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF383
PubMed Links
Gene Details
- Gene
- ZNF383
- Gene Name
- zinc finger protein 383
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152604.1 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
NP_689817.1 |
XM_005258585.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_005258642.1 |
XM_005258587.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_005258644.1 |
XM_005258588.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_005258645.1 |
XM_011526586.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_011524888.1 |
XM_011526587.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_011524889.1 |
XM_011526588.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_011524890.1 |
XM_011526589.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_011524891.1 |
XM_011526590.2 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_011524892.1 |
XM_017026422.1 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_016881911.1 |
XM_017026423.1 |
1435 |
Missense Mutation |
AGT,GGT |
S9G |
XP_016881912.1 |
XM_017026424.1 |
1435 |
Intron |
|
|
XP_016881913.1 |
View Full Product Details