Product Details
- SNP ID
-
rs148830742
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12580690 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGTGTGACTTCTTTCGTGCCTT[C/T]GAAGGTGACTGAAGTAAATGAAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF490
PubMed Links
Gene Details
- Gene
- ZNF490
- Gene Name
- zinc finger protein 490
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020714.2 |
1514 |
Missense Mutation |
CAA,CGA |
Q462R |
NP_065765.1 |
View Full Product Details