Product Details

SNP ID
rs148884848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12351142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGTGTATGTGGCTTCTCTCCA[A/T]ATTCCTGACACTCATCTGGTTTGTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 482 Missense Mutation TAT,TTT Y148F NP_110451.1
XM_006722908.3 482 Missense Mutation TAT,TTT Y141F XP_006722971.1
XM_017027316.1 482 Missense Mutation TAT,TTT Y156F XP_016882805.1
XM_017027317.1 482 Missense Mutation TAT,TTT Y91F XP_016882806.1
XM_017027318.1 482 Missense Mutation TAT,TTT Y148F XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details