Product Details

SNP ID
rs149022147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39707299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACAGTTGTGTGTTTGGCATA[C/T]GGAGATATGAGGAGAAATGCTACTA
Phenotype
MIM: 607260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS14 PubMed Links

Gene Details

Gene
LGALS14
Gene Name
galectin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020129.2 437 Missense Mutation CGG,TGG R72W NP_064514.1
NM_203471.1 437 Missense Mutation CGG,TGG R101W NP_982297.1

View Full Product Details