Product Details

SNP ID
rs149027462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45885016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCATCGAAGGCGAACACTCGCCC[C/T]ACCAGCCCGTGATCCTTCTTGAAGC
Phenotype
MIM: 615331
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP1 PubMed Links

Gene Details

Gene
IRF2BP1
Gene Name
interferon regulatory factor 2 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015649.2 1155 Silent Mutation GTA,GTG V253V NP_056464.1
Gene
MYPOP
Gene Name
Myb related transcription factor, partner of profilin
There are no transcripts associated with this gene.

View Full Product Details