Product Details

SNP ID
rs149133767
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4445544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGGAGCTCGGGTTTCAGGATG[C/G]AGTCCGGCTCGGCCCCCGCGGCCTT
Phenotype
MIM: 601246 MIM: 611946
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CHAF1A PubMed Links

Gene Details

Gene
CHAF1A
Gene Name
chromatin assembly factor 1 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005483.2 1808 Intron NP_005474.2
XM_011527605.2 1808 Intron XP_011525907.1
XM_011527606.2 1808 Intron XP_011525908.1
XM_011527607.2 1808 Intron XP_011525909.1
Gene
MIR4746
Gene Name
microRNA 4746
There are no transcripts associated with this gene.

Gene
UBXN6
Gene Name
UBX domain protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171091.1 1808 Missense Mutation TCC,TGC S374C NP_001164562.1
NM_025241.2 1808 Missense Mutation TCC,TGC S427C NP_079517.1
XM_017027325.1 1808 Missense Mutation TCC,TGC S461C XP_016882814.1

View Full Product Details