Product Details
- SNP ID
-
rs149211453
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12526508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGAAAGCTTTCCCACATTCTTTA[C/T]GTAAGGTTTATCTCCATTATGAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF564
PubMed Links
Gene Details
- Gene
- ZNF564
- Gene Name
- zinc finger protein 564
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144976.3 |
1810 |
Missense Mutation |
ATA,GTA |
I534V |
NP_659413.1 |
View Full Product Details