Product Details

SNP ID
rs149248211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32838194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAAAAGGCCGCTTAATATAAA[C/T]AAGGCTTTATAGGTAGGGTCCTTTA
Phenotype
MIM: 604144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC7A9 PubMed Links

Gene Details

Gene
SLC7A9
Gene Name
solute carrier family 7 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126335.1 Intron NP_001119807.1
NM_001243036.1 Intron NP_001229965.1
NM_014270.4 Intron NP_055085.1
XM_006722992.1 Intron XP_006723055.1
XM_011526402.2 Intron XP_011524704.1
XM_017026230.1 Intron XP_016881719.1
Gene
TDRD12
Gene Name
tudor domain containing 12
There are no transcripts associated with this gene.

View Full Product Details