Product Details

SNP ID
rs149352488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11324150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATCCTACCTCCTCACCCCTTCC[A/G]CCCAACCCCCAGCATCCCTTAGAAG
Phenotype
MIM: 604350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB3D PubMed Links

Gene Details

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004283.3 2169 UTR 3 NP_004274.1
Gene
TSPAN16
Gene Name
tetraspanin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282509.1 2169 Intron NP_001269438.1
NM_001282510.1 2169 Intron NP_001269439.1
NM_012466.3 2169 Intron NP_036598.1
XM_011527901.1 2169 Intron XP_011526203.1
XM_011527902.1 2169 Intron XP_011526204.1
XM_011527903.2 2169 Intron XP_011526205.1

View Full Product Details