Product Details

SNP ID
rs149502504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2071298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTTCATGGAGTTCTTTGAGGG[A/C]TCGGCTGGGAGGTCCCTCTCCGGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MOB3A PubMed Links

Gene Details

Gene
MOB3A
Gene Name
MOB kinase activator 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130807.2 3069 UTR 3 NP_570719.1
XM_011527683.2 3069 UTR 3 XP_011525985.1

View Full Product Details