Product Details

SNP ID
rs149537532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10107832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGCGCCTCCCAGGAGGCCCCA[C/T]CTTGACCTTCCAGGTCAAGAAGGTG
Phenotype
MIM: 609336 MIM: 603913 MIM: 602697 MIM: 607793
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL6 PubMed Links

Gene Details

Gene
ANGPTL6
Gene Name
angiopoietin like 6
There are no transcripts associated with this gene.

Gene
EIF3G
Gene Name
eukaryotic translation initiation factor 3 subunit G
There are no transcripts associated with this gene.

Gene
P2RY11
Gene Name
purinergic receptor P2Y11
There are no transcripts associated with this gene.

Gene
PPAN
Gene Name
peter pan homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020230.5 492 Missense Mutation ACC,ATC T107I NP_064615.3
Gene
PPAN-P2RY11
Gene Name
PPAN-P2RY11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040664.2 492 Missense Mutation ACC,ATC T107I NP_001035754.1
NM_001198690.1 492 Missense Mutation ACC,ATC T107I NP_001185619.1
Gene
SNORD105
Gene Name
small nucleolar RNA, C/D box 105
There are no transcripts associated with this gene.

Gene
SNORD105B
Gene Name
small nucleolar RNA, C/D box 105B
There are no transcripts associated with this gene.

View Full Product Details