Product Details

SNP ID
rs149546342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18434937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGTGGTGGGCATTGGGGGCTC[C/T]TCTTGCAGATGCCCATTGGCATCAC
Phenotype
MIM: 600284 MIM: 611670 MIM: 607391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELL PubMed Links

Gene Details

Gene
ELL
Gene Name
elongation factor for RNA polymerase II
There are no transcripts associated with this gene.

Gene
ISYNA1
Gene Name
inositol-3-phosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170938.1 1575 Silent Mutation GAA,GAG E497E NP_001164409.1
NM_001253389.1 1575 Silent Mutation GAA,GAG E423E NP_001240318.1
NM_016368.4 1575 Silent Mutation GAA,GAG E551E NP_057452.1
XM_011528059.1 1575 Silent Mutation GAA,GAG E619E XP_011526361.1
Gene
SSBP4
Gene Name
single stranded DNA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009998.3 1575 Intron NP_001009998.1
NM_032627.4 1575 Intron NP_116016.1
XM_005259790.3 1575 Intron XP_005259847.1
XM_006722665.3 1575 Intron XP_006722728.1
XM_006722666.2 1575 Intron XP_006722729.1
XM_006722668.2 1575 Intron XP_006722731.1
XM_017026431.1 1575 Intron XP_016881920.1
XM_017026432.1 1575 Intron XP_016881921.1
XM_017026433.1 1575 Intron XP_016881922.1
XM_017026434.1 1575 Intron XP_016881923.1
XM_017026435.1 1575 Intron XP_016881924.1
XM_017026436.1 1575 Intron XP_016881925.1
XM_017026437.1 1575 Intron XP_016881926.1

View Full Product Details