Product Details

SNP ID
rs149600313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41969470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTAACTGTCTTCCAGGTCATTGA[C/T]GGTGCGGTCATCCCAGTTCAGCCGG
Phenotype
MIM: 182350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256213.1 2892 Missense Mutation ATC,GTC I896V NP_001243142.1
NM_001256214.1 2892 Missense Mutation ATC,GTC I898V NP_001243143.1
NM_152296.4 2892 Missense Mutation ATC,GTC I885V NP_689509.1

View Full Product Details