Product Details

SNP ID
rs149604219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21809741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTTTTATATCTTCTCAGTGTC[A/G]CTTTTTGGAAAGGATCTTTTATATG
Phenotype
MIM: 603972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF43 PubMed Links

Gene Details

Gene
ZNF43
Gene Name
zinc finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256648.1 6124 Intron NP_001243577.1
NM_001256649.1 6124 Missense Mutation GCG,GTG A93V NP_001243578.1
NM_001256650.1 6124 Intron NP_001243579.1
NM_001256651.1 6124 Intron NP_001243580.1
NM_001256653.1 6124 Intron NP_001243582.1
NM_001256654.1 6124 Missense Mutation GCG,GTG A34V NP_001243583.1
NM_003423.3 6124 Missense Mutation GCG,GTG A99V NP_003414.2
XM_011528257.2 6124 Intron XP_011526559.1
XM_011528259.2 6124 Missense Mutation GCG,GTG A93V XP_011526561.1
XM_017027207.1 6124 Intron XP_016882696.1
XM_017027208.1 6124 Intron XP_016882697.1
XM_017027209.1 6124 Intron XP_016882698.1
XM_017027210.1 6124 Intron XP_016882699.1
XM_017027211.1 6124 Missense Mutation GCG,GTG A93V XP_016882700.1
XM_017027212.1 6124 Intron XP_016882701.1
XM_017027213.1 6124 Intron XP_016882702.1
XM_017027214.1 6124 Intron XP_016882703.1
XM_017027215.1 6124 Intron XP_016882704.1
XM_017027216.1 6124 Intron XP_016882705.1

View Full Product Details