Product Details

SNP ID
rs149608934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4231252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGCCTCTCGGTACCCGATCGC[C/T]GTGGATTGCTCCTGGACCCTGCCGC
Phenotype
MIM: 605816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD24 PubMed Links

Gene Details

Gene
ANKRD24
Gene Name
ankyrin repeat domain 24
There are no transcripts associated with this gene.

Gene
EBI3
Gene Name
Epstein-Barr virus induced 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005755.2 386 Silent Mutation GCC,GCT A43A NP_005746.2
XM_011527619.2 386 Silent Mutation GCC,GCT A43A XP_011525921.1

View Full Product Details