Product Details

SNP ID
rs149656164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45914547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGTTGGCCCCTGGCGCCCCCAG[C/T]CCCTGATCCTGCCCCAGCGGAGGCC
Phenotype
MIM: 608228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NANOS2 PubMed Links

Gene Details

Gene
NANOS2
Gene Name
nanos C2HC-type zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029861.2 232 Silent Mutation GGA,GGG G49G NP_001025032.1

View Full Product Details