Product Details
- SNP ID
-
rs149713601
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5844602 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCGGGCACCATCTCTGAACAGC[A/G]GGACAGAGCCACAGGGATGTGGAAA
- Phenotype
-
MIM: 111100
MIM: 136836
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FUT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs812936] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUT3
- Gene Name
- fucosyltransferase 3 (Lewis blood group)
- Gene
- FUT6
- Gene Name
- fucosyltransferase 6
There are no transcripts associated with this gene.
- Gene
- LOC101928844
- Gene Name
- uncharacterized LOC101928844
There are no transcripts associated with this gene.
View Full Product Details