Product Details
- SNP ID
-
rs149828471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35500576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTTTCTGCAGTGATGACGCGTCC[A/G]CACCCTGAAAGGAAGAAGGGGCCAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DMKN
PubMed Links
Gene Details
- Gene
- DMKN
- Gene Name
- dermokine
View Full Product Details