Product Details

SNP ID
rs149854108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18899685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGCTCTGGGCCAGCCTCAGGC[A/G]CTGGGAGCGTACTGTAGCACCAGCC
Phenotype
MIM: 606919 MIM: 606942 MIM: 602880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERS1 PubMed Links

Gene Details

Gene
CERS1
Gene Name
ceramide synthase 1
There are no transcripts associated with this gene.

Gene
COPE
Gene Name
coatomer protein complex subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007263.3 950 Silent Mutation AGC,AGT S307S NP_009194.2
NM_199442.1 950 Silent Mutation AGC,AGT S256S NP_955474.1
NM_199444.1 950 Silent Mutation AGC,AGT S255S NP_955476.1
XM_011527655.1 950 Silent Mutation AGC,AGT S330S XP_011525957.1
XM_011527656.1 950 Silent Mutation AGC,AGT S278S XP_011525958.1
Gene
GDF1
Gene Name
growth differentiation factor 1
There are no transcripts associated with this gene.

View Full Product Details