Product Details

SNP ID
rs149917178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4157200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATCCTGCCCAGCCTGGCAAGG[A/G]GCCCTGCCTCTCCTATCATCCTGGC
Phenotype
MIM: 611998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB3L3 PubMed Links

Gene Details

Gene
CREB3L3
Gene Name
cAMP responsive element binding protein 3 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271995.1 509 Missense Mutation GAG,GGG E120G NP_001258924.1
NM_001271996.1 509 Missense Mutation GAG,GGG E121G NP_001258925.1
NM_001271997.1 509 Missense Mutation GAG,GGG E121G NP_001258926.1
NM_032607.2 509 Missense Mutation GAG,GGG E121G NP_115996.1

View Full Product Details