Product Details

SNP ID
rs149951954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53798301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCTCAGCCAGCAACCACAGGC[C/T]CCAGTCTCCCAGGTCGTTGAAACTC
Phenotype
MIM: 611954 MIM: 609648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR373 PubMed Links

Gene Details

Gene
MIR373
Gene Name
microRNA 373
There are no transcripts associated with this gene.

Gene
NLRP12
Gene Name
NLR family pyrin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277126.1 2930 Missense Mutation NP_001264055.1
NM_001277129.1 2930 Intron NP_001264058.1
NM_144687.3 2930 Missense Mutation NP_653288.1
XM_011527479.1 2930 Missense Mutation XP_011525781.1
XM_011527480.1 2930 Intron XP_011525782.1
XM_011527482.1 2930 Intron XP_011525784.1
XM_017027460.1 2930 Missense Mutation XP_016882949.1
XM_017027461.1 2930 Missense Mutation XP_016882950.1
XM_017027462.1 2930 Missense Mutation XP_016882951.1
XM_017027463.1 2930 Missense Mutation XP_016882952.1
XM_017027464.1 2930 Missense Mutation XP_016882953.1
XM_017027465.1 2930 Missense Mutation XP_016882954.1
XM_017027466.1 2930 Missense Mutation XP_016882955.1
XM_017027467.1 2930 Missense Mutation XP_016882956.1

View Full Product Details