Product Details

SNP ID
rs150008188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7526534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTTCCTGCTGGGCTACTCGGA[C/T]GGAGCGGATGACACCTTCGCAGCCT
Phenotype
MIM: 605248 MIM: 603197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCOLN1 PubMed Links

Gene Details

Gene
MCOLN1
Gene Name
mucolipin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020533.2 474 Silent Mutation GAC,GAT D111D NP_065394.1
Gene
PNPLA6
Gene Name
patatin like phospholipase domain containing 6
There are no transcripts associated with this gene.

Gene
ZNF358
Gene Name
zinc finger protein 358
There are no transcripts associated with this gene.

View Full Product Details