Product Details

SNP ID
rs150045306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45765532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGGGCTCCTCCACAGGCACC[A/G]ACTGGAGCTGGGTCAGAACCTTGGC
Phenotype
MIM: 605377 MIM: 600963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHMG1 PubMed Links

Gene Details

Gene
BHMG1
Gene Name
basic helix-loop-helix and HMG-box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310124.1 2584 Intron NP_001297053.1
XM_011527571.2 2584 Intron XP_011525873.1
XM_011527573.2 2584 Intron XP_011525875.1
XM_011527574.2 2584 Intron XP_011525876.1
Gene
DMPK
Gene Name
dystrophia myotonica protein kinase
There are no transcripts associated with this gene.

Gene
SIX5
Gene Name
SIX homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175875.4 2584 Missense Mutation TCG,TTG S730L NP_787071.2

View Full Product Details