Product Details
- SNP ID
-
rs150102707
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14042486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCTGAAGACCATACCCCTGACC[C/T]CTGTTGAGATCCAAGATTTGGAGCT
- Phenotype
-
MIM: 605350
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL27RA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35026308] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL27RA
- Gene Name
- interleukin 27 receptor subunit alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004843.3 |
701 |
Missense Mutation |
CCT,TCT |
P190S |
NP_004834.1 |
View Full Product Details