Product Details
- SNP ID
-
rs150203756
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11868380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGGACTCACACTGGAGAGAAACC[A/G]TATCAATGTAAGGAATGTGGGAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF439
PubMed Links
Gene Details
- Gene
- ZNF439
- Gene Name
- zinc finger protein 439
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152262.2 |
1439 |
Silent Mutation |
CCA,CCG |
P437P |
NP_689475.1 |
View Full Product Details