Product Details

SNP ID
rs150247144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54041788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGGGAGGACTCACCGAGAGAGA[C/T]ACCCTTTCCATATTGGATAAATCTG
Phenotype
MIM: 616804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSTM1 PubMed Links

Gene Details

Gene
VSTM1
Gene Name
V-set and transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288791.1 1065 Silent Mutation GTA,GTG V106V NP_001275720.1
NM_001288792.1 1065 Silent Mutation GTA,GTG V163V NP_001275721.1
NM_001288793.1 1065 Silent Mutation GTA,GTG V74V NP_001275722.1
NM_198481.3 1065 Silent Mutation GTA,GTG V194V NP_940883.2
XM_011526845.1 1065 Silent Mutation GTA,GTG V322V XP_011525147.1
XM_011526846.1 1065 Silent Mutation GTA,GTG V292V XP_011525148.1
XM_011526847.1 1065 Missense Mutation TAT,TGT Y315C XP_011525149.1
XM_011526848.1 1065 Silent Mutation GTA,GTG V261V XP_011525150.1
XM_011526849.1 1065 Silent Mutation GTA,GTG V224V XP_011525151.1
XM_017026666.1 1065 Intron XP_016882155.1

View Full Product Details