Product Details

SNP ID
rs150348108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44751372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCCACCCGTGCAGATGAGGA[C/T]GGAGACACGTGAGTGACAGTCCCCT
Phenotype
MIM: 109560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL3 PubMed Links

Gene Details

Gene
BCL3
Gene Name
B-cell CLL/lymphoma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005178.4 1906 Silent Mutation GAC,GAT D134D NP_005169.2
XM_011527197.2 1906 Silent Mutation GAC,GAT D219D XP_011525499.2
XM_011527198.2 1906 Silent Mutation GAC,GAT D356D XP_011525500.2
XM_011527200.2 1906 Silent Mutation GAC,GAT D38D XP_011525502.1
XM_017027109.1 1906 Silent Mutation GAC,GAT D94D XP_016882598.1
XM_017027110.1 1906 Silent Mutation GAC,GAT D94D XP_016882599.1
Gene
MIR8085
Gene Name
microRNA 8085
There are no transcripts associated with this gene.

View Full Product Details