Product Details

SNP ID
rs150415301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47977419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATCCTTCGTAGGTCTTGTTTAA[C/T]GAGCACCACTTGTGTCTTGCCTTGG
Phenotype
MIM: 612213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSPH1 PubMed Links

Gene Details

Gene
BSPH1
Gene Name
binder of sperm protein homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128326.2 299 Silent Mutation NP_001121798.1
XM_017026118.1 299 Silent Mutation XP_016881607.1

View Full Product Details