Product Details

SNP ID
rs150430942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16479220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCTCTTCCTCCTCTTCCTCGC[A/G]GGCCTTCTTCATTTCCTCCTTGGCC
Phenotype
MIM: 611414 MIM: 616826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALR3 PubMed Links

Gene Details

Gene
CALR3
Gene Name
calreticulin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145046.4 1129 Missense Mutation NP_659483.2
Gene
EPS15L1
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
There are no transcripts associated with this gene.

View Full Product Details