Product Details

SNP ID
rs150515170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50906958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAACTGGCCTGGACGGTTTTCTC[C/T]ATCCACTCAGTGAATTTGCAGAGGT
Phenotype
MIM: 603767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK4 PubMed Links

Gene Details

Gene
KLK4
Gene Name
kallikrein related peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302961.1 741 Missense Mutation ATA,ATG I152M NP_001289890.1
NM_004917.4 741 Missense Mutation ATA,ATG I247M NP_004908.4
XM_005259441.4 741 Missense Mutation ATA,ATG I152M XP_005259498.2
XM_011527545.2 741 Intron XP_011525847.1
XM_011527546.2 741 UTR 3 XP_011525848.1

View Full Product Details