Product Details

SNP ID
rs2302422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54729706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCCCTGTGACCCACGGAGGGA[A/C]CTACAGATGCTTCGGCTCTTTCCGT
Phenotype
MIM: 604938 MIM: 610095
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIR2DL3 PubMed Links

Gene Details

Gene
KIR2DL3
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
There are no transcripts associated with this gene.

Gene
KIR3DL3
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153443.4 919 Missense Mutation AAC,ACC N290T NP_703144.3

View Full Product Details