Product Details

SNP ID
rs150571856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54664259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGGAAAGAGCGTGACCCTGCT[A/G]TGTCAGTCACGGAGCCCAATGGACA
Phenotype
MIM: 604821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRB4 PubMed Links

Gene Details

Gene
LILRB4
Gene Name
leukocyte immunoglobulin like receptor B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278426.3 668 Silent Mutation CTA,CTG L143L NP_001265355.2
NM_001278427.3 668 Silent Mutation CTA,CTG L143L NP_001265356.2
NM_001278428.3 668 Silent Mutation CTA,CTG L143L NP_001265357.2
NM_001278429.3 668 Silent Mutation CTA,CTG L172L NP_001265358.2
NM_001278430.3 668 Intron NP_001265359.2
XM_017026215.1 668 Silent Mutation CTA,CTG L184L XP_016881704.1
XM_017026216.1 668 Silent Mutation CTA,CTG L184L XP_016881705.1
XM_017026217.1 668 Silent Mutation CTA,CTG L184L XP_016881706.1

View Full Product Details