Product Details

SNP ID
rs150594098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:918590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTTCCTCCTGTGCTGCGTCCC[C/T]TTCACGGCCCTGCTGTACCCGCTGC
Phenotype
MIM: 603265 MIM: 604161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID3A PubMed Links

Gene Details

Gene
ARID3A
Gene Name
AT-rich interaction domain 3A
There are no transcripts associated with this gene.

Gene
KISS1R
Gene Name
KISS1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032551.4 1101 Silent Mutation CCC,CCT P97P NP_115940.2
XM_017027382.1 1101 Silent Mutation CCC,CCT P97P XP_016882871.1
Gene
R3HDM4
Gene Name
R3H domain containing 4
There are no transcripts associated with this gene.

View Full Product Details