Product Details

SNP ID
rs150611446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49151304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTCGAGCGACTGGGTCAGGGTT[C/G]CGGCGTTTCCAGCATGTCTGCCAGG
Phenotype
MIM: 142705 MIM: 603144 MIM: 606936
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HRC PubMed Links

Gene Details

Gene
HRC
Gene Name
histidine rich calcium binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002152.2 2279 Missense Mutation CAA,GAA Q698E NP_002143.1
XM_017026733.1 2279 Intron XP_016882222.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 2279 Intron NP_003651.1
XM_017027407.1 2279 Intron XP_016882896.1
Gene
TRPM4
Gene Name
transient receptor potential cation channel subfamily M member 4
There are no transcripts associated with this gene.

View Full Product Details