Product Details

SNP ID
rs150638075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38803880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAAGTACTTACAGGGTAAGGTG[G/T]CATCATCGGGGGTCCCTGTGGGCAC
Phenotype
MIM: 602518
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LGALS4 PubMed Links

Gene Details

Gene
LGALS4
Gene Name
galectin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006149.3 1148 Missense Mutation ACA,CCA T164P NP_006140.1
XM_011526973.1 1148 Missense Mutation ACA,CCA T164P XP_011525275.1
XM_011526974.1 1148 Missense Mutation ACA,CCA T33P XP_011525276.1

View Full Product Details