Product Details

SNP ID
rs150722930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17226294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTCCGACAGCAGATCCAGGCT[C/T]GGAGCTCCAGACGCTGGGACAGGTG
Phenotype
MIM: 132880 MIM: 610675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
There are no transcripts associated with this gene.

Gene
OCEL1
Gene Name
occludin/ELL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024578.2 91 Missense Mutation TCG,TTG S16L NP_078854.1
XM_005260079.2 91 Intron XP_005260136.1
XM_006722899.3 91 Missense Mutation TCG,TTG S16L XP_006722962.1
XM_017027306.1 91 UTR 5 XP_016882795.1
Gene
USE1
Gene Name
unconventional SNARE in the ER 1
There are no transcripts associated with this gene.

View Full Product Details