Product Details

SNP ID
rs150808250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43599104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTTGTCCTGACTGTGGCAAGA[C/T]CTTTGGGCAGGCTGTTTCTCTGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
There are no transcripts associated with this gene.

Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

Gene
ZNF576
Gene Name
zinc finger protein 576
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145347.1 532 Missense Mutation ACC,ATC T120I NP_001138819.1
NM_024327.2 532 Missense Mutation ACC,ATC T120I NP_077303.1

View Full Product Details