Product Details

SNP ID
rs150818557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17324838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCCATCTGGCCGGGCCTGCCC[A/G]CCCGCCCCGTTGGACCCAGGCTCAG
Phenotype
MIM: 610216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO8 PubMed Links

Gene Details

Gene
ANO8
Gene Name
anoctamin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020959.2 3369 Silent Mutation GGC,GGT G1070G NP_066010.1
XM_017027048.1 3369 Silent Mutation GGC,GGT G1070G XP_016882537.1
Gene
DDA1
Gene Name
DET1 and DDB1 associated 1
There are no transcripts associated with this gene.

View Full Product Details