Product Details

SNP ID
rs151153509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38945018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGGACCGGGTCAGGTGAGGCT[G/T]AGAACTTGACCACTTCCTTTTCATA
Phenotype
MIM: 609094
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024907.6 776 Nonsense Mutation TAA,TCA *215S NP_079183.4
NM_148169.2 776 Nonsense Mutation TAA,TCA *224S NP_680474.1

View Full Product Details