Product Details

SNP ID
rs151175533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11806477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCAGTTGGCACAGTTCTGTTC[C/G]AATCCATGAAAGAACTCACACTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
There are no transcripts associated with this gene.

Gene
ZNF491
Gene Name
zinc finger protein 491
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152356.3 750 Missense Mutation CCA,CGA P175R NP_689569.2
XM_005259730.3 750 Missense Mutation CCA,CGA P175R XP_005259787.1

View Full Product Details