Product Details

SNP ID
rs151201212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10274835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCGGGGAGGCTCCGTGCTGGT[A/G]ACATGCAGCACCTCCTGTGACCAGC
Phenotype
MIM: 147840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 457 Silent Mutation GTA,GTG V46V NP_000192.2

View Full Product Details