Product Details

SNP ID
rs151276756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58060003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGTTCCTGCCAGAAGCCAGGGC[A/T]TGACCCCTGGGGTGCGCGTCTCCAC
Phenotype
MIM: 604077
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF135 PubMed Links

Gene Details

Gene
ZNF135
Gene Name
zinc finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164527.1 110 Intron NP_001157999.1
NM_001164529.1 110 Intron NP_001158001.1
NM_001164530.1 110 Intron NP_001158002.1
NM_001289401.1 110 Missense Mutation ATG,TTG M1L NP_001276330.1
NM_001289402.1 110 UTR 5 NP_001276331.1
NM_003436.3 110 Missense Mutation ATG,TTG M1L NP_003427.3
NM_007134.1 110 Intron NP_009065.1
XM_005259211.1 110 Intron XP_005259268.1
XM_006723362.3 110 Missense Mutation ATG,TTG M1L XP_006723425.1
XM_006723363.3 110 Missense Mutation ATG,TTG M1L XP_006723426.1
XM_017027240.1 110 Missense Mutation ATG,TTG M1L XP_016882729.1
XM_017027241.1 110 UTR 5 XP_016882730.1
XM_017027242.1 110 Intron XP_016882731.1
Gene
ZSCAN1
Gene Name
zinc finger and SCAN domain containing 1
There are no transcripts associated with this gene.

View Full Product Details