Product Details
- SNP ID
-
rs151280911
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15425632 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATCTCTCGCAGTGTGTCGATGGG[C/T]GAACCATTGACGGACCACTCGGTCA
- Phenotype
-
MIM: 609475
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP8L
PubMed Links
Gene Details
- Gene
- AKAP8L
- Gene Name
- A-kinase anchoring protein 8 like
There are no transcripts associated with this gene.
- Gene
- WIZ
- Gene Name
- widely interspaced zinc finger motifs
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021241.2 |
2511 |
Silent Mutation |
TCA,TCG |
S406S |
NP_067064.2 |
XM_005260004.4 |
2511 |
Silent Mutation |
TCA,TCG |
S1501S |
XP_005260061.1 |
XM_005260005.4 |
2511 |
Silent Mutation |
TCA,TCG |
S1444S |
XP_005260062.1 |
XM_005260006.4 |
2511 |
Silent Mutation |
TCA,TCG |
S1368S |
XP_005260063.1 |
XM_005260007.4 |
2511 |
Silent Mutation |
TCA,TCG |
S1311S |
XP_005260064.1 |
XM_005260008.2 |
2511 |
Silent Mutation |
TCA,TCG |
S764S |
XP_005260065.1 |
XM_005260009.2 |
2511 |
Silent Mutation |
TCA,TCG |
S580S |
XP_005260066.1 |
XM_005260010.3 |
2511 |
Silent Mutation |
TCA,TCG |
S580S |
XP_005260067.1 |
XM_005260011.2 |
2511 |
Silent Mutation |
TCA,TCG |
S574S |
XP_005260068.1 |
XM_005260012.2 |
2511 |
Silent Mutation |
TCA,TCG |
S447S |
XP_005260069.1 |
XM_006722828.3 |
2511 |
Silent Mutation |
TCA,TCG |
S727S |
XP_006722891.1 |
XM_011528163.2 |
2511 |
Silent Mutation |
TCA,TCG |
S580S |
XP_011526465.1 |
View Full Product Details